3,4-Dimethoxy-5-nitrobenzoic acid 97%
Supplier: Ambeed
|
Warning
|
A269207-100MG
A269207-250MG
77276-734EA
33
USD
77276-734
77276-736
3,4-Dimethoxy-5-nitrobenzoic acid 97%
3,4-Dimethoxy-5-nitrobenzoic acid
Precaución: for Research Use Only
Formula:
C₉H₉NO₆ |
MDL Number:
MFCD00015925 CAS Number: 91004-48-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...