(±)-3-Buten-2-ol 97% stabilized

Supplier: Ambeed
Danger

Synonyms: (±)-Methyl vinyl carbinol, (±)-3-Hydroxy-1-butene, 1-Buten-3-ol

Stabilization: Stabilised with 4-tert-butylpyrocatechol (TBC)

A268984-25G A268984-5G
77276-732EA 75.19 USD
77276-732 77593-664
(±)-3-Buten-2-ol 97% stabilized
(±)-3-Buten-2-ol

Precaución: for Research Use Only

Formula: C₄H₈O
MW: 72.11 g/mol
MDL Number: MFCD00004543
CAS Number: 598-32-3
UN: 1987
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR