Felodipine 98%

Supplier: Ambeed
Warning

A268185-100G A268185-10G A268185-10MG A268185-1MG A268185-25G A268185-50MG A268185-5G
77238-670EA 317 USD
77238-670 77238-672 77238-674 77238-676 77238-678 77238-680 77238-682
Felodipine 98%
Felodipine

Precaución: for Research Use Only

Formula: C₁₈H₁₉Cl₂NO₄
MDL Number: MFCD00868316
CAS Number: 72509-76-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR