2-Fluoro-(4-methoxycarbonyl)phenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Fluoro-4-(methoxycarbonyl)phenylboronic acid, 2-Fluoro-4-(methoxycarbonyl)benzeneboronic acid, 2-Fluoro-4-methoxycarbonylphenylboronic acid

A268145-1G A268145-25G A268145-5G
77262-398EA 14.42 USD
77262-398 77262-400 77262-402
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid 98%
2-Fluoro-(4-methoxycarbonyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₈H₈BFO₄
MW: 197.96 g/mol
MDL Number: MFCD08436034
CAS Number: 603122-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR