2-Chloro-5-nitrobenzenesulfonamide 97%

Supplier: Ambeed
Warning

A268023-100G A268023-10G A268023-1G A268023-25G A268023-500G A268023-5G
77262-386EA 186.43 USD
77262-386 77262-388 77262-390 77262-392 77262-394 77262-396
2-Chloro-5-nitrobenzenesulfonamide 97%
2-Chloro-5-nitrobenzenesulfonamide

Precaución: for Research Use Only

Formula: C₆H₅ClN₂O₄S
MW: 236.63 g/mol
MDL Number: MFCD00035780
CAS Number: 96-72-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR