1,3-Bis[4-(tert-butyl)phenyl]-2,2-Bis[(S)-4-phenyl-4,5-dihydro-2-oxazolyl]propane 98%

Supplier: Ambeed
Warning

Synonyms: (4S,4'S)-2,2'-(1,3-Bis[4-(t-butyl)phenyl)propane-2,2-diyl]bis(4-phenyl-4,5-dihydrooxazole)

A267547-100MG A267547-1G A267547-250MG
77250-270EA 54 USD
77250-270 77250-272 77250-274
1,3-Bis[4-(tert-butyl)phenyl]-2,2-Bis[(S)-4-phenyl-4,5-dihydro-2-oxazolyl]propane 98%
1,3-Bis[4-(tert-butyl)phenyl]-2,2-Bis[(S)-4-phenyl-4,5-dihydro-2-oxazolyl]propane

Precaución: for Research Use Only

Formula: C₄₁H₄₆N₂O₂
MDL Number: MFCD26404519
CAS Number: 1428328-51-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR