6,7-Dimethoxy-4-quinazolone 97%

Supplier: Ambeed
Warning

A267501-100G A267501-10G A267501-1G A267501-25G A267501-5G
77276-706EA 90 USD
77276-706 77276-708 77276-710 77276-712 77276-714
6,7-Dimethoxy-4-quinazolone 97%
6,7-Dimethoxy-4-quinazolone

Precaución: for Research Use Only

Formula: C₁₀H₁₀N₂O₃
MDL Number: MFCD01570147
CAS Number: 13794-72-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR