1H-Imidazole-2(3H)-thione 97%

Supplier: Ambeed
Warning

A267361-250MG A267361-5G A267361-1G
77262-364EA 13.39 USD
77262-364 77262-366 77748-509
1H-Imidazole-2(3H)-thione 97%
1H-Imidazole-2(3H)-thione

Precaución: for Research Use Only

Formula: C₃H₄N₂S
MW: 100.14 g/mol
MDL Number: MFCD00057186
CAS Number: 872-35-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR