IC261 98%

Supplier: Ambeed
Warning

Synonyms: 3-(2,4,6-Ttrimethoxybenzylidene)indolin-2-one, (E)-3-(2,4,6-trimethoxybenzylidene)indolin-2-one, SU-5607

A267194-10MG A267194-1MG A267194-25MG A267194-50MG A267194-5MG
77238-650EA 27.81 USD
77238-650 77238-652 77238-654 77238-656 77238-658
IC261 98%
IC261

Precaución: for Research Use Only

Formula: C₁₈H₁₇NO₄
MW: 311.34 g/mol
MDL Number: MFCD00118156
CAS Number: 186611-52-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR