4-Amino-3-(trifluoromethoxy)benzonitrile 97%

Supplier: Ambeed
Warning

A267188-1G A267188-5G
77238-646EA 38.11 USD
77238-646 77238-648
4-Amino-3-(trifluoromethoxy)benzonitrile 97%
4-Amino-3-(trifluoromethoxy)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₅F₃N₂O
MW: 202.1300 g/mol
MDL Number: MFCD00204177
CAS Number: 175278-23-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR