Derazantinib (ARQ087) 99+%

Supplier: Ambeed
Warning

Synonyms: Derazantinib, (R)-6-(2-Fluorophenyl)-N-(3-(2-((2-methoxyethyl)amino)ethyl)phenyl)-5,6-dihydrobenzo[h]quinazolin-2-amine, ARQ-087, (6R)-6-(2-fluorophenyl)-N-(3-{2-[(2-methoxyethyl)amino]ethyl}phenyl)-5,6-dihydrobenzo[h]quinazolin-2-amine

A267187-1MG A267187-25MG A267187-50MG A267187-5MG A267187-10MG
77276-696EA 27.81 USD
77276-696 77276-698 77276-700 77276-702 77616-128
Derazantinib (ARQ087) 99+%
Derazantinib (ARQ087)

Precaución: for Research Use Only

Formula: C₂₉H₂₉FN₄O
MW: 468.58 g/mol
MDL Number: MFCD30532770
CAS Number: 1234356-69-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR