tert-Butyl 2,5-dichloronicotinate 97%

Supplier: Ambeed
Warning

A267029-1G A267029-250MG A267029-5G
77262-358EA 39.14 USD
77262-358 77262-360 77262-362
tert-Butyl 2,5-dichloronicotinate 97%
tert-Butyl 2,5-dichloronicotinate

Precaución: for Research Use Only

Formula: C₁₀H₁₁Cl₂NO₂
MW: 248.10 g/mol
MDL Number: MFCD30188612
CAS Number: 2089378-26-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR