[4-(Benzyloxy)phenyl]acetylene 98%

Supplier: Ambeed
Warning

Synonyms: 4'-Benzyloxyphenyl acetylene

A266686-1G A266686-250MG A266686-5G
77289-370EA 45.32 USD
77289-370 77289-372 77289-374
[4-(Benzyloxy)phenyl]acetylene 98%
[4-(Benzyloxy)phenyl]acetylene

Precaución: for Research Use Only

Formula: C₁₅H₁₂O
MW: 208.255 g/mol
MDL Number: MFCD01814885
CAS Number: 84284-70-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR