Hydroxy-PEG2-CH2CO2t-Bu 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 2-(2-(2-hydroxyethoxy)ethoxy)acetate, Hydroxy-PEG2-CH2-Boc

A266302-100MG A266302-1G A266302-250MG A266302-5G
77262-346EA 11 USD
77262-346 77262-348 77262-350 77262-352
Hydroxy-PEG2-CH2CO2t-Bu 95%
Hydroxy-PEG2-CH2CO2t-Bu

Precaución: for Research Use Only

Formula: C₁₀H₂₀O₅
MDL Number: MFCD25959245
CAS Number: 149299-82-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR