6-Bromo-3-fluoropyridine-2-carboxaldehyde 98%

Supplier: Ambeed
Warning

A266131-10G A266131-1G A266131-250MG A266131-5G
77276-682EA 131 USD
77276-682 77276-684 77276-686 77276-688
6-Bromo-3-fluoropyridine-2-carboxaldehyde 98%
6-Bromo-3-fluoropyridine-2-carboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₃BrFNO
MDL Number: MFCD07781224
CAS Number: 885267-36-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR