3,4-Dimethoxybenzylamine 98%

Supplier: Ambeed
Danger

A265779-10G A265779-25G A265779-5G
77276-670EA 23 USD
77276-670 77276-672 77276-674
3,4-Dimethoxybenzylamine 98%
3,4-Dimethoxybenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
MDL Number: MFCD00008116
CAS Number: 5763-61-1
UN: 2735
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR