AA26-9 98%

Supplier: Ambeed
Warning

Synonyms: Pyrrolidin-1-yl(1H-1,2,3-triazol-1-yl)methanone, Pyrrolidin-1-yl(triazol-1-yl)methanone

A265641-10MG A265641-1MG A265641-25MG A265641-50MG A265641-5MG
77262-328EA 225.57 USD
77262-328 77262-330 77262-332 77262-334 77262-336
AA26-9 98%
AA26-9

Precaución: for Research Use Only

Formula: C₇H₁₀N₄O
MW: 166.18 g/mol
MDL Number: MFCD28386498
CAS Number: 1312782-34-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR