Orexin 2 98+%

Supplier: Ambeed
Warning

Synonyms: Orexin 2 Receptor Agonist, 4'-methoxy-N,N-dimethyl-3'-(N-(3-((2-(3-methylbenzamido)ethyl)amino)phenyl)sulphamoyl)-[1,1'-biphenyl]-3-carboxamide

A265471-10MG A265471-1MG A265471-2MG A265471-50MG A265471-5MG
77238-622EA 46 USD
77238-622 77238-624 77238-626 77238-628 77238-630
Orexin 2 98+%
Orexin 2

Precaución: for Research Use Only

Formula: C₃₂H₃₄N₄O₅S
MW: 586.22 g/mol
MDL Number: MFCD29924727
CAS Number: 1796565-52-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR