(R)-9-(4-(1-aminopropan-2-yl)phenyl)-8-hydroxy-6-methylthieno[2,3-c]quinolin-4(5H)-one 98%

Supplier: Ambeed
Warning

A265446-10MG A265446-1MG A265446-25MG A265446-50MG A265446-5MG
77238-612EA 72 USD
77238-612 77238-614 77238-616 77238-618 77238-620
(R)-9-(4-(1-aminopropan-2-yl)phenyl)-8-hydroxy-6-methylthieno[2,3-c]quinolin-4(5H)-one 98%
(R)-9-(4-(1-aminopropan-2-yl)phenyl)-8-hydroxy-6-methylthieno[2,3-c]quinolin-4(5H)-one

Precaución: for Research Use Only

Formula: C₂₁H₂₀N₂O₂S
MDL Number: MFCD29924710
CAS Number: 1338540-63-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR