3',4'-Dimethoxyacetophenone 98%

Supplier: Ambeed
Warning

Synonyms: 1-(3,4-Dimethoxyphenyl)ethan-1-one, 3,4-Dimethoxyacetophenone, 1-(3,4-Dimethoxyphenyl)ethanone

A265414-100G A265414-10G A265414-25G A265414-500G A265414-5G
77276-662EA 33.99 USD
77276-662 77276-664 77276-666 77276-668 77748-505
3',4'-Dimethoxyacetophenone 98%
3',4'-Dimethoxyacetophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
MDL Number: MFCD00008737
CAS Number: 1131-62-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR