2-Amino-1-morpholino-1-ethanone hydrochloride 98%

Supplier: Ambeed
Warning

A265287-100MG A265287-250MG A265287-5G
77250-248EA 27 USD
77250-248 77250-250 77568-520
2-Amino-1-morpholino-1-ethanone hydrochloride 98%
2-Amino-1-morpholino-1-ethanone hydrochloride

Precaución: For research use only.

Formula: C₆H₁₃ClN₂O₂
MDL Number: MFCD07366740
CAS Number: 24152-96-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR