(2S,2'S)-1,1'-Bis[(S)-(dimethylamino)phenylmethyl]-2,2'-bis(diphenylphosphino)ferrocene 98%
Supplier: Ambeed
|
Warning
|
A265249-100MG
A265249-250MG
A265249-50MG
A265249-1G
77250-242EA
178
USD
77250-242
77250-244
77250-246
77746-452
(2S,2'S)-1,1'-Bis[(S)-(dimethylamino)phenylmethyl]-2,2'-bis(diphenylphosphino)ferrocene 98%
(2S,2'S)-1,1'-Bis[(S)-(dimethylamino)phenylmethyl]-2,2'-bis(diphenylphosphino)ferrocene
Precaución: for Research Use Only
Formula:
C₅₂H₅₀FeN₂P₂ MW: 820.76 g/mol |
MDL Number:
MFCD08561142 CAS Number: 223725-09-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...