4-Chloro-1,8-naphthalic anhydride 94%

Supplier: Ambeed
Warning

A265057-1000G A265057-500G
77289-306EA 141.11 USD
77289-306 77289-308
4-Chloro-1,8-naphthalic anhydride 94%
4-Chloro-1,8-naphthalic anhydride

Precaución: for Research Use Only

Formula: C₁₂H₅ClO₃
MW: 232.63 g/mol
MDL Number: MFCD00006928
CAS Number: 4053-08-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR