3,5-Di-tert-butylphenol 97%

Supplier: Ambeed
Warning

A264961-100G A264961-10G A264961-1G A264961-500G A264961-5G A264961-25G A264961-250MG
77276-650EA 169 USD
77276-650 77276-652 77276-654 77276-656 77276-658 77616-118 77748-504
3,5-Di-tert-butylphenol 97%
3,5-Di-tert-butylphenol

Precaución: for Research Use Only

Formula: C₁₄H₂₂O
MDL Number: MFCD00008829
CAS Number: 1138-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR