2-Nitro-4-pyridinecarboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 2-Nitroisonicotinic acid

A264725-10G A264725-1G A264725-250MG A264725-5G
77276-640EA 325.48 USD
77276-640 77276-642 77276-644 77276-646
2-Nitro-4-pyridinecarboxylic acid 98%
2-Nitro-4-pyridinecarboxylic acid

Precaución: for Research Use Only

Formula: C₆H₄N₂O₄
MW: 168.11 g/mol
MDL Number: MFCD04114160
CAS Number: 33225-74-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR