TG6-10-1 98%

Supplier: Ambeed
Warning

A264622-10MG A264622-1MG A264622-25MG A264622-50MG A264622-5MG
77238-594EA 32 USD
77238-594 77238-596 77238-598 77238-600 77238-602
TG6-10-1 98%
TG6-10-1

Precaución: for Research Use Only

Formula: C₂₃H₂₃F₃N₂O₄
MDL Number: MFCD29472234
CAS Number: 1415716-58-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR