1-(2-Furoyl)piperazine hydrochloride 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Furan-2-yl(piperazin-1-yl)methanone hydrochloride
A264309-100G
A264309-10G
A264309-1G
A264309-25G
A264309-5G
77250-200EA
316.21
USD
77250-200
77250-202
77250-204
77250-206
77250-208
1-(2-Furoyl)piperazine hydrochloride 95%
1-(2-Furoyl)piperazine hydrochloride
Precaución: for Research Use Only
Formula:
C₉H₁₃ClN₂O₂ MW: 216.6647 g/mol |
MDL Number:
MFCD00236012 CAS Number: 60548-09-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...