4-Isopropylbenzene-1,3-diamine 97%

Supplier: Ambeed
Warning

A264219-100MG A264219-250MG
77238-586EA 48.41 USD
77238-586 77238-588
4-Isopropylbenzene-1,3-diamine 97%
4-Isopropylbenzene-1,3-diamine

Precaución: for Research Use Only

Formula: C₉H₁₄N₂
MW: 150.22 g/mol
MDL Number: MFCD00047849
CAS Number: 14235-45-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR