2-([1,1'-Biphenyl]-4-yl)-5,6-dihydroimidazo[2,1-b]thiazole hydrobromide 98%

Supplier: Ambeed
Warning

A264187-10MG A264187-1MG A264187-25MG A264187-5MG
77276-626EA 134.93 USD
77276-626 77276-628 77276-630 77276-632
2-([1,1'-Biphenyl]-4-yl)-5,6-dihydroimidazo[2,1-b]thiazole hydrobromide 98%
2-([1,1'-Biphenyl]-4-yl)-5,6-dihydroimidazo[2,1-b]thiazole hydrobromide

Precaución: for Research Use Only

Formula: C₁₇H₁₅BrN₂S
MW: 359.28 g/mol
MDL Number: MFCD07281912
CAS Number: 1049780-58-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR