ISCK03 98%

Supplier: Ambeed
Warning

Synonyms: N-(4-(1H-Imidazol-1-yl)phenyl)-4-(tert-butyl)benzenesulphonamide, Stem-Cell Factor/c-Kit inhibitor

A263965-100MG A263965-10MG A263965-1MG A263965-25MG A263965-50MG A263965-5MG
77262-288EA 615.94 USD
77262-288 77262-290 77262-292 77262-294 77262-296 77262-298
ISCK03 98%
ISCK03

Precaución: for Research Use Only

Formula: C₁₉H₂₁N₃O₂S
MW: 355.50 g/mol
MDL Number: MFCD12031591
CAS Number: 945526-43-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR