trans-4-Methyl-β-nitrostyrene 95%

Supplier: Ambeed
Warning

Synonyms: (E)-1-Methyl-4-(2-nitrovinyl)benzene

A263954-25G A263954-5G
77276-616EA 202.91 USD
77276-616 77276-618
trans-4-Methyl-β-nitrostyrene 95%
trans-4-Methyl-β-nitrostyrene

Precaución: for Research Use Only

Formula: C₉H₉NO₂
MW: 163.18 g/mol
MDL Number: MFCD00024828
CAS Number: 7559-36-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR