Piceid 98%

Supplier: Ambeed
Warning

Synonyms: Resveratrol 3-Glucoside, (E)-Piceid, 4',5-Dihydroxystilben-3-yl beta-D-Glucopyranoside, Polydatin, trans-Resveratrol-3-O-β-D-Glucopyranoside, trans-Polydatin, (E)-Polydatin, 3,4,5-Trihydroxystilbene-3-beta-monoglucoside, 3,4,5-Tsg, (2S,3R,4S,5S,6R)-2-(3-Hydroxy-5-((E)-4-hydroxystyryl)phenoxy)-6-(hydroxymethyl)tetrahydro-2H-pyran-3,4,5-triol

A263851-100MG A263851-1G A263851-250MG A263851-25G A263851-5G A263851-100G
77289-280EA 5 USD
77289-280 77289-282 77289-284 77289-286 77289-288 77748-502
Piceid 98%
Piceid

Precaución: for Research Use Only

Formula: C₂₀H₂₂O₈
MDL Number: MFCD00210592
CAS Number: 27208-80-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR