N-(2-Chloro-6-methylphenyl)-2-((6-(4-(2-hydroxyethyl)piperazin-1-yl)-2-methylpyrimidin-4-yl)amino)thiazole-5-carboxamide hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: Dasatinib HCl

A263536-1MG A263536-50MG A263536-5MG A263536-100MG A263536-250MG
77238-580EA 15.45 USD
77238-580 77238-582 77616-112 77746-449 77746-450
N-(2-Chloro-6-methylphenyl)-2-((6-(4-(2-hydroxyethyl)piperazin-1-yl)-2-methylpyrimidin-4-yl)amino)thiazole-5-carboxamide hydrochloride 98%
N-(2-Chloro-6-methylphenyl)-2-((6-(4-(2-hydroxyethyl)piperazin-1-yl)-2-methylpyrimidin-4-yl)amino)thiazole-5-carboxamide hydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₂₇Cl₂N₇O₂S
MW: 524.47 g/mol
MDL Number: MFCD26960525
CAS Number: 854001-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR