4-Nitrophenyl palmitate 98%

Supplier: Ambeed
Warning

Synonyms: 4-Nitrophenyl hexadecanoate

A263494-5G A263494-1G A263494-25G
77289-270EA 49.44 USD
77289-270 77616-110 77748-500
4-Nitrophenyl palmitate 98%
4-Nitrophenyl palmitate

Precaución: for Research Use Only

Formula: C₂₂H₃₅NO₄
MW: 377.52 g/mol
MDL Number: MFCD00047732
CAS Number: 1492-30-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR