2-Chloro-4-(trifluoromethyl)benzoic acid 97%

Supplier: Ambeed
Warning

Synonyms: 2-Chloro-4-trifluoromethylbenzoic acid, 2-Chloro-α,α,α-trifluoro-p-toluic acid

A263483-10G A263483-1G A263483-250MG A263483-25G A263483-5G
77262-276EA 37 USD
77262-276 77262-278 77262-280 77262-282 77262-284
2-Chloro-4-(trifluoromethyl)benzoic acid 97%
2-Chloro-4-(trifluoromethyl)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClF₃O₂
MDL Number: MFCD06208178
CAS Number: 23228-45-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR