N-(4-Bromo-2-fluorophenyl)-6-methoxy-7-((1-methylpiperidin-4-yl)methoxy)quinazolin-4-amine hydrochloride 98+%

Supplier: Ambeed
Warning

A263359-25MG A263359-5MG
77238-562EA 5.15 USD
77238-562 77238-564 77238-566
N-(4-Bromo-2-fluorophenyl)-6-methoxy-7-((1-methylpiperidin-4-yl)methoxy)quinazolin-4-amine hydrochloride 98+%
N-(4-Bromo-2-fluorophenyl)-6-methoxy-7-((1-methylpiperidin-4-yl)methoxy)quinazolin-4-amine hydrochloride

Precaución: for Research Use Only

Formula: C₂₂H₂₅BrClFN₄O₂
CAS Number: 524722-52-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR