2,5-Dibromo-4-fluorotoluene 97%

Supplier: Ambeed
Warning

A263286-100G A263286-1G A263286-25G A263286-5G
77262-268EA 325.01 USD
77262-268 77262-270 77262-272 77262-274
2,5-Dibromo-4-fluorotoluene 97%
2,5-Dibromo-4-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅Br₂F
MDL Number: MFCD00044738
CAS Number: 134332-29-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR