4'-(4-Bromophenyl)acetophenone 98%

Supplier: Ambeed
Warning

A263271-100G A263271-1G A263271-25G
77289-264EA 308 USD
77289-264 77289-266 77289-268
4'-(4-Bromophenyl)acetophenone 98%
4'-(4-Bromophenyl)acetophenone

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrO
MDL Number: MFCD00143242
CAS Number: 5731-01-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR