2-Amino-2-[3-(trifluoromethyl)phenyl]acetic acid 97%

Supplier: Ambeed
Warning

Synonyms: 3-(Trifluoromethyl)phenylglycine, H-DL-Phg(3-CF₃)-OH, 2-Amino-2-(3-(trifluoromethyl)phenyl)acetic acid

A263267-10G A263267-1G A263267-250MG A263267-5G
77262-260EA 113.3 USD
77262-260 77262-262 77262-264 77262-266
2-Amino-2-[3-(trifluoromethyl)phenyl]acetic acid 97%
2-Amino-2-[3-(trifluoromethyl)phenyl]acetic acid

Precaución: for Research Use Only

Formula: C₉H₈F₃NO₂
MW: 219.16 g/mol
MDL Number: MFCD02662404
CAS Number: 242475-26-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR