3-Amino-4-nitrobenzoic acid 95%

Supplier: Ambeed
Warning

A263155-1G A263155-5G
77276-600EA 59.74 USD
77276-600 77276-602
3-Amino-4-nitrobenzoic acid 95%
3-Amino-4-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₆N₂O₄
MDL Number: MFCD01806270
CAS Number: 6968-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR