4,4'-Dinitro-[1,1'-biphenyl]-2,2'-dicarboxylic acid 97%

Supplier: Ambeed
Danger

A262933-250MG A262933-5G
77238-538EA 7.21 USD
77238-538 77238-540
4,4'-Dinitro-[1,1'-biphenyl]-2,2'-dicarboxylic acid 97%
4,4'-Dinitro-[1,1'-biphenyl]-2,2'-dicarboxylic acid

Precaución: for Research Use Only

Formula: C₁₄H₈N₂O₈
MW: 332.22 g/mol
MDL Number: MFCD21337356
CAS Number: 20246-81-5
UN: 1325
ADR: 4.1,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR