3-Fluoro-D-phenylalanine 97%

Supplier: Ambeed
Warning

Synonyms: H-D-Phe(3-F)-OH

A262797-1G A262797-250MG A262797-5G A262797-100G A262797-10G
77250-190EA 14.42 USD
77250-190 77250-192 77250-194 77746-441 77746-442
3-Fluoro-D-phenylalanine 97%
3-Fluoro-D-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MW: 183.18 g/mol
MDL Number: MFCD00066449
CAS Number: 110117-84-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR