3-Fluoro-DL-phenylalanine 95%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-3-(3-fluorophenyl)propionic acid, H-DL-Phe(3-F)-OH

A262287-100G A262287-10G A262287-1G A262287-250MG A262287-25G A262287-5G
77250-178EA 325 USD
77250-178 77250-180 77250-182 77250-184 77250-186 77250-188
3-Fluoro-DL-phenylalanine 95%
3-Fluoro-DL-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₁₀FNO₂
MDL Number: MFCD00004273
CAS Number: 2629-54-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR