3-Amino-1,2,4-triazine 97%

Supplier: Ambeed
Warning

A262181-10G A262181-1G A262181-25G A262181-500G A262181-5G A262181-100G
77276-572EA 9 USD
77276-572 77276-574 77276-576 77276-578 77276-580 77616-100
3-Amino-1,2,4-triazine 97%
3-Amino-1,2,4-triazine

Precaución: for Research Use Only

Formula: C₃H₄N₄
MDL Number: MFCD00006459
CAS Number: 1120-99-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR