2,2'-Bis[(4S)-4-Benzyl-2-Oxazoline] ≥98%, ee 99%

Supplier: Ambeed
Warning

A261993-100MG A261993-1G A261993-50MG A261993-5G A261993-250MG
77250-156EA 19 USD
77250-156 77250-158 77250-160 77250-162 77550-646
2,2'-Bis[(4S)-4-Benzyl-2-Oxazoline] ≥98%, ee 99%
2,2'-Bis[(4S)-4-Benzyl-2-Oxazoline]

Precaución: for Research Use Only

Formula: C₂₀H₂₀N₂O₂
MDL Number: MFCD00191793
CAS Number: 133463-88-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR