trans-1-Ethoxy-4-(4-n-propycyclohexyl)benzene 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
1-Ethoxy-4-(trans-4-propylcyclohexyl)benzene
A261984-1G
A261984-25G
A261984-5G
77289-228EA
6.18
USD
77289-228
77289-230
77289-232
trans-1-Ethoxy-4-(4-n-propycyclohexyl)benzene 98%
trans-1-Ethoxy-4-(4-n-propycyclohexyl)benzene
Precaución: for Research Use Only
Formula:
C₁₇H₂₆O MW: 246.394 g/mol |
MDL Number:
MFCD06658195 CAS Number: 80944-44-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...