1-Fluoro-2,4,6-trimethylpyridiniumtetrafluoroborate 98%

Supplier: Ambeed
Danger

A261822-250MG A261822-25G
77238-520EA 37 USD
77238-520 77238-522
1-Fluoro-2,4,6-trimethylpyridiniumtetrafluoroborate 98%
1-Fluoro-2,4,6-trimethylpyridinium tetrafluoroborate

Precaución: for Research Use Only

Formula: C₈H₁₁BF₅N
MDL Number: MFCD00153182
CAS Number: 109705-14-8
UN: 3260
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR