2-Methoxy-3-methylbenzoic acid 95%

Supplier: Ambeed
Warning

A261783-1G A261783-250MG A261783-5G
77262-240EA 81.37 USD
77262-240 77262-242 77262-244
2-Methoxy-3-methylbenzoic acid 95%
2-Methoxy-3-methylbenzoic acid

Precaución: for Research Use Only

Formula: C₉H₁₀O₃
MDL Number: MFCD00016522
CAS Number: 26507-91-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR