KC7F2 98%

Supplier: Ambeed
Warning

Synonyms: N,N'-(Disulphanediylbis(ethane-2,1-diyl))bis(2,5-dichlorobenzenesulphonamide)

A261687-10MG A261687-1MG A261687-25MG A261687-50MG A261687-5MG
77238-510EA 56.65 USD
77238-510 77238-512 77238-514 77238-516 77238-518
KC7F2 98%
KC7F2

Precaución: for Research Use Only

Formula: C₁₆H₁₆Cl₄N₂O₄S₄
MW: 570.38 g/mol
MDL Number: MFCD22683809
CAS Number: 927822-86-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR