1,3,4-Thiadiazole-2,5-diamine 95%

Supplier: Ambeed
Warning

A261247-1G A261247-10G
77250-154EA 19 USD
77250-154 77746-432
1,3,4-Thiadiazole-2,5-diamine 95%
1,3,4-Thiadiazole-2,5-diamine

Precaución: for Research Use Only

Formula: C₂H₄N₄S
MDL Number: MFCD00043511
CAS Number: 2937-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR